missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenoxyacetonitrile, 97%, Thermo Scientific™

Catalog Number p-7045377
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
103140-12-1
C8H5Cl2NO
202.03
MFCD00052707
PNROREDTZJCOHF-UHFFFAOYSA-N
2-3,5-dichlorophenoxy acetonitrile, 3,5-dichlorophenoxyacetonitrile, 2-3,5-dichlorophenoxy ethanenitrile, maybridge1_008353, acmc-1bu1u, 3,5-dichlorophenoxy acetonitrile, acetonitrile,2-3,5-dichlorophenoxy
517810
2-(3,5-dichlorophenoxy)acetonitrile
ClC1=CC(OCC#N)=CC(Cl)=C1
This item is not returnable. View return policy
103140-12-1
C8H5Cl2NO
202.03
MFCD00052707
PNROREDTZJCOHF-UHFFFAOYSA-N
2-3,5-dichlorophenoxy acetonitrile, 3,5-dichlorophenoxyacetonitrile, 2-3,5-dichlorophenoxy ethanenitrile, maybridge1_008353, acmc-1bu1u, 3,5-dichlorophenoxy acetonitrile, acetonitrile,2-3,5-dichlorophenoxy
517810
2-(3,5-dichlorophenoxy)acetonitrile
ClC1=CC(OCC#N)=CC(Cl)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 60°C
Quantity 1 g
UN Number UN3439
Beilstein 4983677
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorophenoxyacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'