missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenol, 99%, Thermo Scientific Chemicals

Catalog Number p-3736189
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
591-35-5
C6H4Cl2O
163
MFCD00002259
VPOMSPZBQMDLTM-UHFFFAOYSA-N
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
11571
3,5-dichlorophenol
C1=C(C=C(C=C1Cl)Cl)O
591-35-5
C6H4Cl2O
163
MFCD00002259
VPOMSPZBQMDLTM-UHFFFAOYSA-N
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624
11571
3,5-dichlorophenol
C1=C(C=C(C=C1Cl)Cl)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 68°C
Color Yellow to Brown
Boiling Point 233°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3OH
Quantity 10 g
Beilstein 06, 190
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,dmso
Formula Weight 163
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material 3, 5-Dichlorophenol
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dichlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'