Learn More
Thermo Scientific Maybridge 3,5-Dichlorobenzylamine, 10g, 95%
Brand: Thermo Scientific Maybridge TL00168EA
3,5-Dichlorobenzylamine, 10g, 95%
Code : 31
Additional Details : CAS Number : 39989-43-0 Weight : 0.06000kg Transport : UN number : 2735 Chem class : 8 Pack group : III