missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzyl alcohol, 98%, Thermo Scientific™

Catalog Number 10760691
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
60211-57-6
C7H6Cl2O
177.03
MFCD00004634
VSNNLLQKDRCKCB-UHFFFAOYSA-N
3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908
43236
(3,5-dichlorophenyl)methanol
C1=C(C=C(C=C1Cl)Cl)CO
This item is not returnable. View return policy
60211-57-6
C7H6Cl2O
177.03
MFCD00004634
VSNNLLQKDRCKCB-UHFFFAOYSA-N
3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908
43236
(3,5-dichlorophenyl)methanol
C1=C(C=C(C=C1Cl)Cl)CO
CAS Min % 97.5
CAS Max % 100.0
Melting Point 76°C to 80°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CH2OH
Quantity 25 g
Formula Weight 177.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dichlorobenzyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dichlorobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'