missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzenesulfonamide, 97%, Thermo Scientific™

Catalog Number p-7037234
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
19797-32-1
C6H5Cl2NO2S
226.071
MFCD00117161
AHNOVNYOUPQVRX-UHFFFAOYSA-N
3,5-dichlorobenzene-1-sulfonamide, 3,5-dichlorobenzenesulphonamide, benzenesulfonamide, 3,5-dichloro, 3,5-bis chloranyl benzenesulfonamide, pubchem11789, maybridge1_001366, acmc-209f2d, spr_4, benzenesulfonamide,3,5-dichloro, 3,5-dichlorobenzenesulfonamide
705607
3,5-dichlorobenzenesulfonamide
C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)N
This item is not returnable. View return policy
19797-32-1
C6H5Cl2NO2S
226.071
MFCD00117161
AHNOVNYOUPQVRX-UHFFFAOYSA-N
3,5-dichlorobenzene-1-sulfonamide, 3,5-dichlorobenzenesulphonamide, benzenesulfonamide, 3,5-dichloro, 3,5-bis chloranyl benzenesulfonamide, pubchem11789, maybridge1_001366, acmc-209f2d, spr_4, benzenesulfonamide,3,5-dichloro, 3,5-dichlorobenzenesulfonamide
705607
3,5-dichlorobenzenesulfonamide
C1=C(C=C(C=C1Cl)Cl)S(=O)(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 161°C to 165°C
Quantity 250 mg
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'