missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichloro-N-methylbenzylamine, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 190.07 |
Percent Purity | 95% |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- 3,5-Dichloro-N-methylbenzylamine
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'