missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichloro-4-hydroxybenzenesulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed in the convergent syntheses involving hydroxyarenesulfonyl chlorides.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed in the convergent syntheses involving hydroxyarenesulfonyl chlorides.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 118°C to 119°C |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 2806977 |
Sensitivity | Moisture sensitive |
Synonym | 3,5-dichloro-4-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-4-hydroxybenzenesulphonyl chloride, 3,5-dichloro-4-hydroxy benzene sulphonyl chloride, 3,5-dichloro-4-hydroxy-benzenesulfonyl chloride, acmc-1c20j, 3,5-dichloro-4-hydroxyphenyl chlorosulfone, 3,5-dichloro-4-hydroxybenzenesulphonylchloride, benzenesulfonylchloride,3,5-dichloro-4-hydroxy, benzenesulfonyl chloride, 3,5-dichloro-4-hydroxy, benzenesulfonylchloride, 3,5-dichloro-4-hydroxy |
Solubility Information | Hydrolyzes in water. |
IUPAC Name | 3,5-dichloro-4-hydroxybenzenesulfonyl chloride |
Formula Weight | 261.51 |
Percent Purity | 97% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'