missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Dichloro-4-fluoro-1-nitrobenzene, 95%

Brand:  Alfa Aesar™ H61321.22

Code : NEW

Additional Details : CAS Number : 3107-19-5 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15447848

  • £325.00 / 100g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Dichloro-4-fluoro-1-nitrobenzene
209.99
3,5-Dichloro-4-fluoronitrobenzene
3107-19-5
MFCD00129963

Specifications

3,5-Dichloro-4-fluoro-1-nitrobenzene
100g
Powder
209.99
3,5-Dichloro-4-fluoronitrobenzene
White
3107-19-5
95%
MFCD00129963
Videos
SDS