missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2,4,6-trifluoropyridine, 98%, ACROS Organics™

£18.91 - £70.44

Chemical Identifiers

CAS 1737-93-5
Molecular Formula C5Cl2F3N
Molecular Weight (g/mol) 201.96
InChI Key PKSORSNCSXBXOT-UHFFFAOYSA-N
Synonym 2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
PubChem CID 61273
IUPAC Name 3,5-dichloro-2,4,6-trifluoropyridine
SMILES C1(=C(C(=NC(=C1Cl)F)F)Cl)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10003630
SDS View Documents
Acros Organics
311400050
5g £18.91
5g View alternative products
Save Up to 
»
Estimated Shipment
04-09-2020
Please to purchase this item. Need a web account? Register with us today!
12397093
SDS View Documents
Acros Organics
311400250
25g £70.44
25g View alternative products
Save Up to 
»
Estimated Shipment
04-09-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1737-93-5
201.96
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
3,5-dichloro-2,4,6-trifluoropyridine
C5Cl2F3N
PKSORSNCSXBXOT-UHFFFAOYSA-N
61273
C1(=C(C(=NC(=C1Cl)F)F)Cl)F

Specifications

3,5-Dichloro-2,4,6-trifluoropyridine
113°C
1.622
24.0°C to 26.0°C
1737-93-5
C5Cl2F3N
PKSORSNCSXBXOT-UHFFFAOYSA-N
3,5-dichloro-2,4,6-trifluoropyridine
61273
98%
1.6220g/mL
Glass bottle
159.0°C to 160.0°C
5g
97.5% min. (GC)
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
C1(=C(C(=NC(=C1Cl)F)F)Cl)F
201.96
201.96