missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-2,4,6-trifluoropyridine, 97+%, Thermo Scientific Chemicals

Catalog Number p-7080612
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1737-93-5
C5Cl2F3N
201.957
MFCD00006226
PKSORSNCSXBXOT-UHFFFAOYSA-N
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
61273
3,5-dichloro-2,4,6-trifluoropyridine
C1(=C(C(=NC(=C1Cl)F)F)Cl)F
This item is not returnable. View return policy
1737-93-5
C5Cl2F3N
201.957
MFCD00006226
PKSORSNCSXBXOT-UHFFFAOYSA-N
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
61273
3,5-dichloro-2,4,6-trifluoropyridine
C1(=C(C(=NC(=C1Cl)F)F)Cl)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 24°C to 26°C
Density 1.648
Boiling Point 159°C to 160°C
Flash Point None
Refractive Index 1.478
Quantity 5 g
UN Number UN2811
Beilstein 475874
Formula Weight 201.96
Percent Purity ≥97%
Chemical Name or Material 3,5-Dichloro-2,4,6-trifluoropyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'