missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Dichloro-1H-indazole, 95%

Brand:  Alfa Aesar™ H33220.03

Code : NEW

Additional Details : CAS Number : 36760-20-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15456027

  • £204.10 / 1g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Description

Description

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3,5-Dichloro-1H-indazole
36760-20-0
187
MFCD09263200

Specifications

3,5-Dichloro-1H-indazole
240° to 242°C
36760-20-0
95%
MFCD09263200
White to cream
1g
Solid
187
Videos
SDS