missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081062
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
625-92-3
C5H3Br2N
236.89
MFCD00014634
SOSPMXMEOFGPIM-UHFFFAOYSA-N
pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln
69369
CHEBI:51593
3,5-dibromopyridine
BrC1=CC(Br)=CN=C1
This item is not returnable. View return policy
625-92-3
C5H3Br2N
236.89
MFCD00014634
SOSPMXMEOFGPIM-UHFFFAOYSA-N
pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln
69369
CHEBI:51593
3,5-dibromopyridine
BrC1=CC(Br)=CN=C1

3-Acetylamino-5-ethoxypyridine was prepared by converting 3, 5-dibromopyridine with sodium ethylate into 3-bromo-5-ethoxypyridine, allowing this substance to interact with ammonia and acetylating the aminoethoxypyridine thus formed with acetic anhydride. Lithiation of 3, 5-dibromopyridine with LDA and subsequent reaction with electrophiles provided 4-alkyl-3, 5-dibromopyridines 2 in high yield. Ligand was prepared by a Pd(0)-catalyzed cross-coupling reaction of 3,5-dibromopyridine and 5-tributylstannyl-3,3?-bipyridine. The synthesis of 3, 5-bis (2-indolyl) pyridine and 3-[(2-indolyl)-5-phenyl] pyridine derivatives as includes Stille or Suzuki type reactions, which were realized on the 3,5-dibromopyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Acetylamino-5-ethoxypyridine was prepared by converting 3, 5-dibromopyridine with sodium ethylate into 3-bromo-5-ethoxypyridine, allowing this substance to interact with ammonia and acetylating the aminoethoxypyridine thus formed with acetic anhydride. Lithiation of 3, 5-dibromopyridine with LDA and subsequent reaction with electrophiles provided 4-alkyl-3, 5-dibromopyridines 2 in high yield. Ligand was prepared by a Pd(0)-catalyzed cross-coupling reaction of 3,5-dibromopyridine and 5-tributylstannyl-3,3′-bipyridine. The synthesis of 3, 5-bis (2-indolyl) pyridine and 3-[(2-indolyl)-5-phenyl] pyridine derivatives as includes Stille or Suzuki type reactions, which were realized on the 3,5-dibromopyridine.

Solubility
Soluble in Chloroform and Methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alkali metals, moisture.
TRUSTED_SUSTAINABILITY
Melting Point 110°C to 113°C
Quantity 5 g
Beilstein 108477
Solubility Information Soluble in Chloroform and Methanol. Insoluble in water.
Formula Weight 236.9
Percent Purity ≥98%
Chemical Name or Material 3,5-Dibromopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'