missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromopyridin-2-amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | Br2(C5H2N)NH2 |
Quantity | 10 g |
Formula Weight | 251.91 |
Percent Purity | 97% |
Chemical Name or Material | 3, 5-Dibromopyridin-2-amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'