missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzyl bromide, 99%, Thermo Scientific™

Catalog Number p-3211801
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
56908-88-4
C7H5Br3
328.83
MFCD00052415
PWTFRUXTAFBWBW-UHFFFAOYSA-N
3,5-dibromobenzyl bromide, 1,3-dibromo-5-bromomethyl benzene, benzene, 1,3-dibromo-5-bromomethyl, alpha,3,5-tribromotoluene, acmc-20andg, 3,5-dibromobenzylbromide, 3,5-dibromo benzyl bromide, 1,3-bis bromanyl-5-bromomethyl benzene
143427
1,3-dibromo-5-(bromomethyl)benzene
BrCC1=CC(Br)=CC(Br)=C1
This item is not returnable. View return policy
56908-88-4
C7H5Br3
328.83
MFCD00052415
PWTFRUXTAFBWBW-UHFFFAOYSA-N
3,5-dibromobenzyl bromide, 1,3-dibromo-5-bromomethyl benzene, benzene, 1,3-dibromo-5-bromomethyl, alpha,3,5-tribromotoluene, acmc-20andg, 3,5-dibromobenzylbromide, 3,5-dibromo benzyl bromide, 1,3-bis bromanyl-5-bromomethyl benzene
143427
1,3-dibromo-5-(bromomethyl)benzene
BrCC1=CC(Br)=CC(Br)=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 93°C to 97°C
Color Beige-Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3CH2Br
Quantity 5 g
Formula Weight 328.83
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 5-Dibromobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dibromobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'