missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-dibromobenzamide, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 25 g |
Formula Weight | 278.93 |
Percent Purity | 97% |
Chemical Name or Material | 3, 5-Dibromobenzamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'