missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzaldehyde +98%, ACROS Organics™

Brand:  Acros Organics 393160250

Code : 29

Additional Details : CAS Number : 609-85-8 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10509613

  • £86.60 / 25g


Estimated Shipment
13-03-2018
Add to basket


Specifications

Chemical Identifiers

3, 5-Dibromoanthranilic acid
98.0
≥98.0%
C7H5Br2NO2
MFCD00017087
609-85-8
100.0
98+%
294.93

Specifications

3, 5-Dibromoanthranilic acid
98.0
Authentic
Powder
White to beige
25g
294.93
609-85-8
100.0
Glass bottle
≥98.0%
98+%
C7H5Br2NO2
MFCD00017087
Videos
SDS