missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Dibromo-4-iodotoluene, 98%

Brand:  Alfa Aesar™ A10502.06

Code : NEW

Additional Details : CAS Number : 175278-10-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11434280

  • £62.80 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Dibromo-4-iodotoluene
C7H5Br2I
MFCD00052397
175278-10-1
375.83
2,6-Dibromo-4-methyliodobenzene

Specifications

3,5-Dibromo-4-iodotoluene
69° to 72°C
98%
C7H5Br2I
MFCD00052397
175278-10-1
126° to 128°C (0.4mmHg)
5g
375.83
2,6-Dibromo-4-methyliodobenzene
Videos
SDS