missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-2-pyridinol, 98%, ACROS Organics™

Brand:  Acros Organics 379670010

Code : 29

Additional Details : CAS Number : 13472-81-6 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10506743

  • £16.50 / 1g


Estimated Shipment
14-02-2018
Add to basket


Specifications

Chemical Identifiers

3, 5-Dibromo-2-pyridinol
97.5
≥97.5%
C5H3Br2NO
MFCD00023472
13472-81-6
100.0
98%
252.89
3, 5-Dibromo-2-hydroxypyridine

Specifications

3, 5-Dibromo-2-pyridinol
204° to 210°C
13472-81-6
97.5
Glass bottle
≥97.5%
C5H3Br2NO
MFCD00023472
Beige to light brown
1g
Authentic
100.0
Crystalline Powder
98%
252.89
3, 5-Dibromo-2-hydroxypyridine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,5-Dibromo-2-pyridinol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS