missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-2-pyridinol, 98%, Thermo Scientific™

Catalog Number p-3211798
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13472-81-6
MFCD00023472
XIFRODWVHSZAMM-UHFFFAOYSA-N
3,5-dibromo-2-hydroxypyridine, 3,5-dibromopyridin-2-ol, 3,5-dibromo-2-pyridinol, 3,5-dibromo-2 1h-pyridinone, 2-hydroxy-3,5-dibromopyridine, 3,5-dibromo-2-pyridone, 3,5-dibromopyridine-2-ol, 2 1h-pyridinone, 3,5-dibromo, pubchem6448, acmc-209bwt
642826
3,5-dibromo-1H-pyridin-2-one
C1=C(C(=O)NC=C1Br)Br
This item is not returnable. View return policy
13472-81-6
MFCD00023472
XIFRODWVHSZAMM-UHFFFAOYSA-N
3,5-dibromo-2-hydroxypyridine, 3,5-dibromopyridin-2-ol, 3,5-dibromo-2-pyridinol, 3,5-dibromo-2 1h-pyridinone, 2-hydroxy-3,5-dibromopyridine, 3,5-dibromo-2-pyridone, 3,5-dibromopyridine-2-ol, 2 1h-pyridinone, 3,5-dibromo, pubchem6448, acmc-209bwt
642826
3,5-dibromo-1H-pyridin-2-one
C1=C(C(=O)NC=C1Br)Br
CAS Min % 97.5
CAS Max % 100.0
Melting Point 204°C to 210°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H3Br2NO
Quantity 1 g
Molecular Weight (g/mol) 252.893
Formula Weight 252.89
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dibromo-2-pyridinol
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dibromo-2-pyridinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'