missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-2-chloropyridine, 98%, Thermo Scientific™

Catalog Number 15418068
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
40360-47-2
C5H2Br2ClN
271.336
MFCD00233991
PYSICVOJSJMFKP-UHFFFAOYSA-N
2-chloro-3,5-dibromopyridine, 3,5-dibromo-2-chloro-pyridine, pubchem6101, acmc-1ar8p, ksc235m9d, tpc-py111, 2-chloro-3,5 dibromo pyridine, pyridine, 3,5-dibromo-2-chloro
817092
3,5-dibromo-2-chloropyridine
C1=C(C=NC(=C1Br)Cl)Br
This item is not returnable. View return policy
40360-47-2
C5H2Br2ClN
271.336
MFCD00233991
PYSICVOJSJMFKP-UHFFFAOYSA-N
2-chloro-3,5-dibromopyridine, 3,5-dibromo-2-chloro-pyridine, pubchem6101, acmc-1ar8p, ksc235m9d, tpc-py111, 2-chloro-3,5 dibromo pyridine, pyridine, 3,5-dibromo-2-chloro
817092
3,5-dibromo-2-chloropyridine
C1=C(C=NC(=C1Br)Cl)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Formula Weight 271.34
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 3,5-Dibromo-2-chloropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'