missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diaminobenzoic Acid Dihydrochloride 99%, ACROS Organics™

Brand:  Acros Organics 151191000

Code : 29

Additional Details : CAS Number : 618-56-4 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10255670

  • £44.20 / 100g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

3, 5-Diaminobenzoic acid dihydrochloride
618-56-4
100.0
99%
(H2N)2C6H3CO2H·2HCl
MFCD00012983
99%
98.5
99%
C7H8N2O2·2HCl
225.08

Specifications

3, 5-Diaminobenzoic acid dihydrochloride
> 300.0°C
99%
98.5
Authentic
Solubility in water: 0.1 g/mL (20°C).
99%
C7H8N2O2·2HCl
225.08
14, 453
Beige to gray
100g
618-56-4
100.0
Glass bottle
Fluffy Fine Crystalline Powder
99%
(H2N)2C6H3CO2H·2HCl
MFCD00012983
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,5-Diaminobenzoic acid dihydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS