missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diaminobenzoic Acid Dihydrochloride 99%, ACROS Organics™

Brand:  Acros Organics 151190100

Code : 29

Additional Details : CAS Number : 618-56-4 Weight : 0.06000kg


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10127953

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3, 5-Diaminobenzoic acid dihydrochloride
618-56-4
100.0
99%
(H2N)2C6H3CO2H·2HCl
MFCD00012983
99%
98.5
99%
C7H8N2O2·2HCl
225.08

Specifications

3, 5-Diaminobenzoic acid dihydrochloride
> 300.0°C
99%
98.5
Authentic
Solubility in water: 0.1 g/mL (20°C).
99%
C7H8N2O2·2HCl
225.08
14, 453
Beige to gray
10g
618-56-4
100.0
Glass bottle
Fluffy Fine Crystalline Powder
99%
(H2N)2C6H3CO2H·2HCl
MFCD00012983
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,5-Diaminobenzoic acid dihydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS