missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diaminobenzoic acid dihydrochloride, 99%, ACROS Organics™
£17.46 - £42.50
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10127953
|
Acros Organics
151190100 |
10g | Glass bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
10021820
|
Acros Organics
151190250 |
25g | Glass bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
10255670
|
Acros Organics
151191000 |
100g | Glass bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3,5-Diaminobenzoic acid dihydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.