missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(trifluoromethyl)phenyl isothiocyanate, ≥97%, Thermo Scientific™

Catalog Number p-3485387
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
23165-29-9
C9H3F6NS
271.18
MFCD00040838
FXOSSGVJGGNASE-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl phenylisothiocyanate, isothiocyanic acid 3,5-bis trifluoromethyl phenyl ester, 3,5-di trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-3,5-bis trifluoromethyl, pubchem5019, acmc-209g1w
2733395
1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F
This item is not returnable. View return policy
23165-29-9
C9H3F6NS
271.18
MFCD00040838
FXOSSGVJGGNASE-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl phenylisothiocyanate, isothiocyanic acid 3,5-bis trifluoromethyl phenyl ester, 3,5-di trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-3,5-bis trifluoromethyl, pubchem5019, acmc-209g1w
2733395
1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F
CAS Min % 97.0
Density 1.4850g/mL
Assay Percent Range 97+%
Linear Formula (CF3)2C6H3NCS
Quantity 10 g
Formula Weight 271.18
Percent Purity 97%
Chemical Name or Material 3, 5-Di(trifluoromethyl)phenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'