3,5-Di-tert-butyl-o-benzoquinone, 97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: BTB10297DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 3383-21-9 Weight: 0.05100kg


Quantity 1g

Product Code. 10352433

Quantity Price
1 £ 8.0 / 1g
Estimated Shipment date
from Supplier 05-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
CAS 3383-21-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H20O2
Linear Formula [(CH3)3C]2C6H2(=O)2
Formula Weight 220.31
MDL Number MFCD00001647

Specification

Formula Weight 220.31
Quantity 1g
Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
Assay 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS Min % 96.5
Assay Percent Range 97%
Linear Formula [(CH3)3C]2C6H2(=O)2
MDL Number MFCD00001647
CAS Max % 100.0
Molecular Formula C14H20O2
CAS 3383-21-9
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.