missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%, Thermo Scientific Chemicals
22.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
Specifications
Melting Point | ∼195°C (decomposition) |
Quantity | 10 g |
Beilstein | 3144594 |
Solubility Information | Soluble in water. |
Formula Weight | 276.37 |
Percent Purity | 98% |
Chemical Name or Material | 3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'