missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Bromo-pyridin-3-yl)-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester, ≥97%, Thermo Scientific™

Catalog Number p-3110374
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
850375-34-7
C10H8BrN3O3
298.096
MFCD06797495
LSHJXSGSPMZMFQ-UHFFFAOYSA-N
ethyl 3-5-bromopyridin-3-yl-1,2,4-oxadiazole-5-carboxylate, 3-5-bromo-pyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester, 1,2,4-oxadiazole-5-carboxylicacid, 3-5-bromo-3-pyridinyl-, ethyl ester, 3-5-bromopyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester
7170018
ethyl 3-(5-bromopyridin-3-yl)-1,2,4-oxadiazole-5-carboxylate
CCOC(=O)C1=NC(=NO1)C2=CC(=CN=C2)Br
This item is not returnable. View return policy
850375-34-7
C10H8BrN3O3
298.096
MFCD06797495
LSHJXSGSPMZMFQ-UHFFFAOYSA-N
ethyl 3-5-bromopyridin-3-yl-1,2,4-oxadiazole-5-carboxylate, 3-5-bromo-pyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester, 1,2,4-oxadiazole-5-carboxylicacid, 3-5-bromo-3-pyridinyl-, ethyl ester, 3-5-bromopyridin-3-yl-1,2,4 oxadiazole-5-carboxylic acid ethyl ester
7170018
ethyl 3-(5-bromopyridin-3-yl)-1,2,4-oxadiazole-5-carboxylate
CCOC(=O)C1=NC(=NO1)C2=CC(=CN=C2)Br
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 298.1
Percent Purity 95%
Chemical Name or Material 3-(5-Bromo-pyridin-3-yl)-[1, 2, 4]oxadiazole-5-carboxylic acid ethyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'