missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals

Catalog Number p-7080600
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
349-58-6
C8H4F6O
230.109
MFCD00000386
ODSXJQYJADZFJX-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
67680
3,5-bis(trifluoromethyl)phenol
C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F
This item is not returnable. View return policy
349-58-6
C8H4F6O
230.109
MFCD00000386
ODSXJQYJADZFJX-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr
67680
3,5-bis(trifluoromethyl)phenol
C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 20°C to 21°C
Density 1.511
Boiling Point 97°C to 98°C (50 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 1 g
Beilstein 2123670
Formula Weight 230.11
Percent Purity 97%
Chemical Name or Material 3,5-Bis (trifluoromethyl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'