missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Bis(trifluoromethyl)cinnamonitrile, 98%

Brand:  Alfa Aesar™ A11279.06

Code : NEW

Additional Details : CAS Number : 175136-63-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11493634

  • £156.03 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Bis (trifluoromethyl)cinnamonitrile
C11H5F6N
MFCD00051605
175136-63-7
265.16
3-[3,5-Bis(trifluoromethyl)phenyl]acrylonitrile

Specifications

3,5-Bis (trifluoromethyl)cinnamonitrile
98%
265.16
3-[3,5-Bis(trifluoromethyl)phenyl]acrylonitrile
5g
175136-63-7
C11H5F6N
MFCD00051605
80° to 83°C
Videos
SDS