missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzylamine, 95%, Thermo Scientific Chemicals

Catalog Number p-3756946
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
85068-29-7
C9H7F6N
243.15
MFCD00009909
DHVHORCFFOSRBP-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzylamine, 3,5-bis trifluoromethyl phenyl methanamine, 3,5-di trifluoromethyl benzylamine, 3,5-ditrifluoromethylbenzylamine, mbt-bym, 3,5-bis trifluoromethyl phenyl methylamine, benzenemethanamine, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzylamine
521099
[3,5-bis(trifluoromethyl)phenyl]methanamine
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN
This item is not returnable. View return policy
85068-29-7
C9H7F6N
243.15
MFCD00009909
DHVHORCFFOSRBP-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzylamine, 3,5-bis trifluoromethyl phenyl methanamine, 3,5-di trifluoromethyl benzylamine, 3,5-ditrifluoromethylbenzylamine, mbt-bym, 3,5-bis trifluoromethyl phenyl methylamine, benzenemethanamine, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzylamine
521099
[3,5-bis(trifluoromethyl)phenyl]methanamine
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 46°C to 55°C
Color White to Beige
Boiling Point 82°C to 84°C (15.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CH2NH2
Quantity 1 g
Formula Weight 243.15
Percent Purity 95%
Physical Form Crystalline Mass, Chunks or Crystals and Powder
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'