missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzamidoxime, 97%

Brand:  Alfa Aesar™ H64759.06

Code : NEW

Additional Details : CAS Number : 72111-09-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15402099

  • £142.04 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Bis(trifluoromethyl)benzamidoxime
72111-09-2
272.15
MFCD00068188

Specifications

3,5-Bis(trifluoromethyl)benzamidoxime
Solid
272.15
129° to 132°C (lit.)
72111-09-2
97%
MFCD00068188
5g
Videos
SDS