missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 10657061
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
401-95-6
C9H4F6O
242.12
MFCD00010206
LDWLIXZSDPXYDR-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j
136247
3,5-bis(trifluoromethyl)benzaldehyde
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
401-95-6
C9H4F6O
242.12
MFCD00010206
LDWLIXZSDPXYDR-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j
136247
3,5-bis(trifluoromethyl)benzaldehyde
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 37°C (1.3 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CHO
Refractive Index 1.4215 to 1.4235
Quantity 5 g
Specific Gravity 1.47
Formula Weight 242.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'