missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)aniline, 98+%, Thermo Scientific Chemicals

Catalog Number 10453965
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
328-74-5
C8H5F6N
229.12
MFCD00000394
CDIDGWDGQGVCIB-UHFFFAOYSA-N
3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl
9480
3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F
This item is not returnable. View return policy
328-74-5
C8H5F6N
229.12
MFCD00000394
CDIDGWDGQGVCIB-UHFFFAOYSA-N
3,5-bis trifluoromethyl aniline, 3,5-di trifluoromethyl aniline, benzenamine, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenamine, 3',5'-bis-trifluoromethyl aniline, 3,5-bis-trifluoromethylaniline, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylidine, 3,5-bis-trifluoromethyl-phenylamine, aniline, 3,5-bis trifluoromethyl
9480
3,5-bis(trifluoromethyl)aniline
C1=C(C=C(C=C1C(F)(F)F)N)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.4700g/mL
Boiling Point 85°C (15.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CF3)2C6H3NH2
Refractive Index 1.4330 to 1.435
Quantity 250 mL
Specific Gravity 1.47
Formula Weight 229.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)aniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'