missing translation for 'onlineSavingsMsg'
Learn More

3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine, 97%, Thermo Scientific™

Catalog Number 10545184
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
845885-86-1
C10H14BNO2
191.04
MFCD03425979
HDQULWCYQQMIJJ-UHFFFAOYSA-N
3-5,5-dimethyl-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid neopentylglycol ester, pyridine-3-boronic acid neopentylglycol ester, pyridine-3-boronic acid, neopentyl ester, pyridine-3-boronic acid neopentyl glycol ester, 2-3-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, 5,5-dimethyl-2-pyridin-3-yl-1,3,2-dioxaborinane, 2-4-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, pyridine-3-boronic acid neopentyl ester, pyridine-3-boronic acid, neopentylglycol ester
2794717
3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
CC1(C)COB(OC1)C1=CC=NC=C1
This item is not returnable. View return policy
845885-86-1
C10H14BNO2
191.04
MFCD03425979
HDQULWCYQQMIJJ-UHFFFAOYSA-N
3-5,5-dimethyl-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid neopentylglycol ester, pyridine-3-boronic acid neopentylglycol ester, pyridine-3-boronic acid, neopentyl ester, pyridine-3-boronic acid neopentyl glycol ester, 2-3-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, 5,5-dimethyl-2-pyridin-3-yl-1,3,2-dioxaborinane, 2-4-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, pyridine-3-boronic acid neopentyl ester, pyridine-3-boronic acid, neopentylglycol ester
2794717
3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
CC1(C)COB(OC1)C1=CC=NC=C1
CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 191.04
Percent Purity 97%
Chemical Name or Material 3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'