missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Thiazolyl)-L-alanine, 95%, Thermo Scientific Chemicals

Catalog Number p-7055033
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
119433-80-6
C6H8N2O2S
172.202
MFCD00079677
WBZIGVCQRXJYQD-YFKPBYRVSA-N
3-4-thiazolyl-l-alanine, s-2-amino-3-thiazol-4-yl propanoic acid, thiazolylalanine, 3-4-thiazoyl-l-alanine, unii-vtl1y3s44b, 2s-2-amino-3-1,3-thiazol-4-yl propanoic acid, vtl1y3s44b, s-3-4-thiazolyl alanine, l-thiazolylalanine, thiazolylalanine, l
2761495
(2S)-2-amino-3-(1,3-thiazol-4-yl)propanoic acid
C1=C(N=CS1)CC(C(=O)O)N
This item is not returnable. View return policy
119433-80-6
C6H8N2O2S
172.202
MFCD00079677
WBZIGVCQRXJYQD-YFKPBYRVSA-N
3-4-thiazolyl-l-alanine, s-2-amino-3-thiazol-4-yl propanoic acid, thiazolylalanine, 3-4-thiazoyl-l-alanine, unii-vtl1y3s44b, 2s-2-amino-3-1,3-thiazol-4-yl propanoic acid, vtl1y3s44b, s-3-4-thiazolyl alanine, l-thiazolylalanine, thiazolylalanine, l
2761495
(2S)-2-amino-3-(1,3-thiazol-4-yl)propanoic acid
C1=C(N=CS1)CC(C(=O)O)N

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store at (-20°C). Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Solubility Information Slightly soluble in water.
Formula Weight 172.2
Percent Purity 95%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material 3-(4-Thiazolyl)-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'