missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Pyridyl)acrylic acid, 97%, ACROS Organics™

£25.52 - £77.86

Chemical Identifiers

CAS 5337-79-1
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00040746
InChI Key SSAYTINUCCRGDR-OWOJBTEDSA-N
Synonym 3-4-pyridyl acrylic acid, trans-3-4-pyridyl acrylic acid, 3-pyridin-4-yl-acrylic acid, 4-pyridineacrylic acid, e-3-pyridin-4-yl acrylic acid, 3-pyridin-4-yl acrylic acid, 2e-3-pyridin-4-ylacrylic acid, 3-4-pyridine acrylic acid, 2e-3-pyridin-4-yl prop-2-enoic acid, e-3-pyridin-4-yl-acrylic acid
PubChem CID 736782
IUPAC Name (E)-3-pyridin-4-ylprop-2-enoic acid
SMILES C1=CN=CC=C1C=CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10025223
SDS View Documents
Acros Organics
391750010
1g Glass bottle
£25.52
1g
View alternative products
Save Up to 
»
Estimated Shipment
08-02-2021
Please to purchase this item. Need a web account? Register with us today!
10177203
SDS View Documents
Acros Organics
391750050
5g Glass bottle
£77.86
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

5337-79-1
149.15
SSAYTINUCCRGDR-OWOJBTEDSA-N
736782
C1=CN=CC=C1C=CC(=O)O
C8H7NO2
MFCD00040746
3-4-pyridyl acrylic acid, trans-3-4-pyridyl acrylic acid, 3-pyridin-4-yl-acrylic acid, 4-pyridineacrylic acid, e-3-pyridin-4-yl acrylic acid, 3-pyridin-4-yl acrylic acid, 2e-3-pyridin-4-ylacrylic acid, 3-4-pyridine acrylic acid, 2e-3-pyridin-4-yl prop-2-enoic acid, e-3-pyridin-4-yl-acrylic acid
(E)-3-pyridin-4-ylprop-2-enoic acid

Specifications

3-(4-Pyridyl)acrylic acid
Glass bottle
280°C
5337-79-1
100.0
C8H7NO2
3-4-pyridyl acrylic acid, trans-3-4-pyridyl acrylic acid, 3-pyridin-4-yl-acrylic acid, 4-pyridineacrylic acid, e-3-pyridin-4-yl acrylic acid, 3-pyridin-4-yl acrylic acid, 2e-3-pyridin-4-ylacrylic acid, 3-4-pyridine acrylic acid, 2e-3-pyridin-4-yl prop-2-enoic acid, e-3-pyridin-4-yl-acrylic acid
C1=CN=CC=C1C=CC(=O)O
149.15
149.15
97%
Authentic
Yellow
1g
96.0
97%
MFCD00040746
SSAYTINUCCRGDR-OWOJBTEDSA-N
(E)-3-pyridin-4-ylprop-2-enoic acid
736782
Crystalline Powder
Safety and Handling

Safety and Handling

Product Identifier
  • 3-(4-Pyridyl)acrylic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS