missing translation for 'onlineSavingsMsg'
Learn More

3,4-Pyridinedicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-189405
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
490-11-9
C7H5NO4
167.12
MFCD00006392
MUYSADWCWFFZKR-UHFFFAOYSA-N
3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine
10273
CHEBI:46860
pyridine-3,4-dicarboxylic acid
OC(=O)C1=CC=NC=C1C(O)=O
This item is not returnable. View return policy
490-11-9
C7H5NO4
167.12
MFCD00006392
MUYSADWCWFFZKR-UHFFFAOYSA-N
3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine
10273
CHEBI:46860
pyridine-3,4-dicarboxylic acid
OC(=O)C1=CC=NC=C1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 262°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 22, 155
Merck Index 15, 2286
Solubility Information Solubility in water: .. Other solubilities: sparingly soluble in hot alcohol,ether and benzen,insoluble in chloroform
Formula Weight 167.12
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3, 4-Pyridinedicarboxylic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Pyridinedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'