missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Oxo-2-thioxo-3-thiazolidinyl)benzoic acid, 95%

Brand:  Alfa Aesar™ H63979.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448616

  • £128.26 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Oxo-2-thioxo-3-thiazolidinyl)benzoic acid
253.29
MFCD01184188

Specifications

3-(4-Oxo-2-thioxo-3-thiazolidinyl)benzoic acid
95%
MFCD01184188
Solid
253.29
1g
Videos
SDS