missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Nitrophenyl)propanoic acid, 98%, ACROS Organics™

Brand:  Acros Organics 458201000

Code : NEW

Additional Details : CAS Number : 16642-79-8 Weight : 0.10000kg


Quantity 100g

 View more versions of this product

Product Code. 15402242

  • £379.48 / 100g

Estimated Shipment: 13-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

16642-79-8
195.17
VZOPVJNBOQOLPN-UHFFFAOYSA-N
85526
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O
C9H9NO4
MFCD00126834
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
3-(4-nitrophenyl)propanoic acid

Specifications

3-(4-Nitrophenyl)propanoic acid
166.0°C to 168.0°C
16642-79-8
C9H9NO4
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O
195.17
195.18
Authentic
100g
97.5% min. (HPLC)
MFCD00126834
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-(4-nitrophenyl)propanoic acid
85526
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-(4-Nitrophenyl)propanoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

SDS Documents
Certificates