missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinylmethyl)indole, 95%, Thermo Scientific™

Catalog Number 15460029
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
5379-88-4
C13H16N2O
216.28
MFCD00458413
VHGISLOQNDQFIW-UHFFFAOYSA-N
3-morpholinomethyl indole, indole, 3-morpholinomethyl, 3-morpholin-4-ylmethyl-1h-indole, 4-27-00-00551 beilstein handbook reference, 4-indol-3-ylmethyl morpholine, 3-morpholinomethyl-1h-indole, morpholine, n-3-indolyl methyl, 4-1h-indol-3-ylmethyl morpholine, 3-4-morpholinylmethyl indole
21477
4-(1H-indol-3-ylmethyl)morpholine
C(N1CCOCC1)C1=CNC2=CC=CC=C12
This item is not returnable. View return policy
5379-88-4
C13H16N2O
216.28
MFCD00458413
VHGISLOQNDQFIW-UHFFFAOYSA-N
3-morpholinomethyl indole, indole, 3-morpholinomethyl, 3-morpholin-4-ylmethyl-1h-indole, 4-27-00-00551 beilstein handbook reference, 4-indol-3-ylmethyl morpholine, 3-morpholinomethyl-1h-indole, morpholine, n-3-indolyl methyl, 4-1h-indol-3-ylmethyl morpholine, 3-4-morpholinylmethyl indole
21477
4-(1H-indol-3-ylmethyl)morpholine
C(N1CCOCC1)C1=CNC2=CC=CC=C12

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Beilstein 193005
Formula Weight 216.28
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(4-Morpholinylmethyl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'