missing translation for 'onlineSavingsMsg'
Learn More
Learn More
{3-[(4-methylpiperidino)methyl]phenyl}methanamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 218.34 |
Percent Purity | 97% |
Chemical Name or Material | {3-[(4-Methylpiperidino)methyl]phenyl}methanamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'