missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde, ≥95%, Maybridge™

£68.31 - £484.00

Chemical Identifiers

CAS 859850-88-7
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
MDL Number MFCD08271925
InChI Key KKYYCUZQPBFXKD-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl methyl benzaldehyde, benzaldehyde,3-4-methyl-1-piperazinyl methyl, 3-4-methylpiperazinyl methyl benzaldehyde, 3-4-methyl-1-piperazinyl methyl benzaldehyde
PubChem CID 18525774
IUPAC Name 3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
SMILES CN1CCN(CC1)CC2=CC=CC(=C2)C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10011104
SDS View Documents
Maybridge
CC45304CB
250mg
£68.31
250mg
View alternative products
Save Up to 
»
Estimated Shipment
05-01-2021
Please to purchase this item. Need a web account? Register with us today!
10597274
SDS View Documents
Maybridge
CC45304DA
1g
£173.00
1g
View alternative products
Save Up to 
»
Estimated Shipment
05-01-2021
Please to purchase this item. Need a web account? Register with us today!
10265794
SDS View Documents
Maybridge
CC45304DE
5g
£484.00
5g
View alternative products
Save Up to 
»
Estimated Shipment
05-01-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

859850-88-7
218.3
KKYYCUZQPBFXKD-UHFFFAOYSA-N
18525774
CN1CCN(CC1)CC2=CC=CC(=C2)C=O
C13H18N2O
MFCD08271925
3-4-methylpiperazin-1-yl methyl benzaldehyde, benzaldehyde,3-4-methyl-1-piperazinyl methyl, 3-4-methylpiperazinyl methyl benzaldehyde, 3-4-methyl-1-piperazinyl methyl benzaldehyde
3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde

Specifications

3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde
250mg
94.5
95%
MFCD08271925
KKYYCUZQPBFXKD-UHFFFAOYSA-N
3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
18525774
95%
Amber Glass Bottle
859850-88-7
100.0
C13H18N2O
3-4-methylpiperazin-1-yl methyl benzaldehyde, benzaldehyde,3-4-methyl-1-piperazinyl methyl, 3-4-methylpiperazinyl methyl benzaldehyde, 3-4-methyl-1-piperazinyl methyl benzaldehyde
CN1CCN(CC1)CC2=CC=CC(=C2)C=O
218.3
218.3
SDS