missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]benzylamine, 95%, Maybridge

Brand:  Maybridge CC55913DA

Code : 31

Additional Details : CAS Number : 915707-48-1 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10472674

  • £190.00 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

3-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]benzylamine
94.5
95%
C14H23N3
MFCD09787493
915707-48-1
100.0
95%
233.36

Specifications

3-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]benzylamine
915707-48-1
100.0
95%
C14H23N3
MFCD09787493
1g
94.5
Amber Glass Bottle
95%
233.36
Videos
SDS