missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.
Solubility
Soluble in water (partly), ethanol, and methanol.
Notes
Store at room temperature. Store away from oxidizing agents. Store in a cool, dry place. Store in a tightly closed container.
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.
Solubility
Soluble in water (partly), ethanol, and methanol.
Notes
Store at room temperature. Store away from oxidizing agents. Store in a cool, dry place. Store in a tightly closed container.
Specifications
Melting Point | 127°C to 130°C |
Quantity | 5 g |
Beilstein | 177739 |
Solubility Information | Soluble in water (partly),ethanol,and methanol. |
Formula Weight | 180.16 |
Percent Purity | 98% |
Chemical Name or Material | 3,4- (Methylenedioxy)phenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'