missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Methyl-1-piperazinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63047.06

Code : NEW

Additional Details : CAS Number : 5444-91-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15489498

  • £478.06 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Methyl-1-piperazinylmethyl)indole
229.32
885083
5444-91-7
MFCD00450888

Specifications

3-(4-Methyl-1-piperazinylmethyl)indole
5444-91-7
95%
MFCD00450888
5g
Solid
229.32
885083
Videos
SDS