missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Methyl-1-piperazinylmethyl)-5-nitroindole, 95%

Brand:  Alfa Aesar™ H63196.03

Code : NEW

Additional Details : CAS Number : 3414-71-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468166

  • £95.61 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Methyl-1-piperazinylmethyl)-5-nitroindole
274.32
1592954
3414-71-9
MFCD01718687

Specifications

3-(4-Methyl-1-piperazinylmethyl)-5-nitroindole
3414-71-9
95%
MFCD01718687
1g
Solid
274.32
1592954
Videos
SDS