missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7030505
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
883738-38-3
C18H27BN2O3
330.24
MFCD05864308
ZYDMNNQZEUSDGG-UHFFFAOYSA-N
4-methylpiperazin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 3-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzamide, 3-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb046
2760029
CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
883738-38-3
C18H27BN2O3
330.24
MFCD05864308
ZYDMNNQZEUSDGG-UHFFFAOYSA-N
4-methylpiperazin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 3-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzamide, 3-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb046
2760029
CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 86°C to 89°C
Quantity 1 g
IUPAC Name (4-methylpiperazin-1-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanone
Formula Weight 330.23
Percent Purity 97%
Chemical Name or Material 3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'