missing translation for 'onlineSavingsMsg'
Learn More

3-(4-hydroxyphenyl)propanohydrazide, Thermo Scientific™

Catalog Number p-4391369
Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy
65330-63-4
C9H12N2O2
180.21
MFCD00025134
JMNHUOPRGLTNJI-UHFFFAOYSA-N
3-4-hydroxyphenyl propanohydrazide, 3-4-hydroxyphenyl propanehydrazide, 3-4-hydroxyphenyl propionohydrazide, benzenepropanoic acid,4-hydroxy-, hydrazide, benzenepropanoic acid, 4-hydroxy-, hydrazide
2796587
3-(4-hydroxyphenyl)propanehydrazide
NNC(=O)CCC1=CC=C(O)C=C1
This item is not returnable. View return policy
65330-63-4
C9H12N2O2
180.21
MFCD00025134
JMNHUOPRGLTNJI-UHFFFAOYSA-N
3-4-hydroxyphenyl propanohydrazide, 3-4-hydroxyphenyl propanehydrazide, 3-4-hydroxyphenyl propionohydrazide, benzenepropanoic acid,4-hydroxy-, hydrazide, benzenepropanoic acid, 4-hydroxy-, hydrazide
2796587
3-(4-hydroxyphenyl)propanehydrazide
NNC(=O)CCC1=CC=C(O)C=C1
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 180.21
Percent Purity 97%
Chemical Name or Material 3-(4-hydroxyphenyl)propanohydrazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'