missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 97%, Thermo Scientific Chemicals
28.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep away from oxidizing agents.
It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Melting Point | 87°C to 92°C |
Quantity | 1 g |
Beilstein | 2110370 |
Solubility Information | Soluble in dimethyl sulfoxide and methanol. |
Formula Weight | 196.2 |
Percent Purity | 97% |
Chemical Name or Material | 3- (4-Hydroxy-3-methoxyphenyl)propionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'