missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-5-methyl-4-isoxazolecarboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2699106
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1736-21-6
C11H8FNO3
221.19
MFCD03407356
PDEGBONVUJDOFN-UHFFFAOYSA-N
3-4-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-4-fluorophenyl-5-methyl, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methylisoxazol-4-carboxylic acid, buttpark 9912-50, 4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methyl, 4-isoxazolecarboxylicacid,3-4-fluorophenyl-5-methyl, 3-4-fluoro-phenyl-5-methyl-isoxazole-4-carboxylic acid
2780709
3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
CC1=C(C(O)=O)C(=NO1)C1=CC=C(F)C=C1
This item is not returnable. View return policy
1736-21-6
C11H8FNO3
221.19
MFCD03407356
PDEGBONVUJDOFN-UHFFFAOYSA-N
3-4-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-4-fluorophenyl-5-methyl, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methylisoxazol-4-carboxylic acid, buttpark 9912-50, 4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methyl, 4-isoxazolecarboxylicacid,3-4-fluorophenyl-5-methyl, 3-4-fluoro-phenyl-5-methyl-isoxazole-4-carboxylic acid
2780709
3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
CC1=C(C(O)=O)C(=NO1)C1=CC=C(F)C=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.19
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorophenyl)-5-methyl-4-isoxazolecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'